Invitrogen
Manufacturer Code:PA5105074
Catalog # PIPA5105074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SMURF1 around the phosphorylation site of Ser200. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4930431E10Rik E3 ubiquitin ligase SMURF1 E3 ubiquitin-protein ligase SMURF1 HECT-type E3 ubiquitin transferase SMURF1 hSMURF1 hSMURF2 KIAA1625 mKIAA1625 RGD1309707 SMAD specific E3 ubiquitin protein ligase 1 Smad ubiquitination regulatory factor 1 Smad-specific E3 ubiquitin ligase 1 SMAD-specific E3 ubiquitin-protein ligase 1 Smurf1; E3 ubiquitin ligase SMURF1; E3 ubiquitin-protein ligase SMURF1; HECT-type E3 ubiquitin transferase SMURF1; hSMURF1; SMAD ubiquitination regulatory factor 1; Smad-specific E3 ubiquitin ligase 1; SMAD-specific E3 ubiquitin-protein ligase 1
Gene Aliases: 4930431E10Rik; KIAA1625; mKIAA1625; RGD1309707; SMURF1
UniProt ID: (Human) Q9HCE7, (Mouse) Q9CUN6
Entrez Gene ID: (Human) 57154, (Rat) 690516, (Mouse) 75788
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.