Invitrogen
Manufacturer Code:700047
Catalog # PI700047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 31H14L11 |
Immunogen | A peptide corresponding to amino acids 456-465 of Q15797 and Q99717. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: BSP-1; JV4-1; JV5-1; MAD homolog 1; MAD homolog 5; MAD, mothers against decapentaplegic homolog 1; MAD, mothers against decapentaplegic homolog 5; Mad-related protein 1; Mothers against decapentaplegic homolog 1; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; mothers against DPP homolog 1; SMA- and MAD-related protein 5; SMAD 1; SMAD 5; SMAD family member 1; SMAD family member 5; SMAD, mothers against DPP homolog 1; SMAD, mothers against DPP homolog 5; SMAD1 SMAD5 BSP-1 MAD homolog 1 MAD mothers against decapentaplegic homolog 1 Mad-related protein 1 TGF-beta signaling protein 1 hSMAD1 mothers against DPP homolog 1 transforming growth factor-beta signaling protein 1 SMAD1 DWFC JV5-1 MADH5 SMAD5; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Aliases: BSP-1; BSP1; DWFC; JV4-1; JV41; JV5-1; MADH1; MADH5; MADR1; SMAD1; SMAD5
UniProt ID: (Human) Q15797, (Human) Q99717
Entrez Gene ID: (Human) 4086, (Human) 4090
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.