Invitrogen
Manufacturer Code:PA595680
Catalog # PIPA595680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr276 conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-hydroxytryptamine (serotonin) transporter; 5HT transporter; 5HTT; serotonin transporter; SERT; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; Solute carrier family 6 member 4; solute carrier family 6, member 4
Gene Aliases: 5-HTT; AI323329; Htt; Sert; Slc6a4
UniProt ID: (Rat) P31652, (Mouse) Q60857
Entrez Gene ID: (Rat) 25553, (Mouse) 15567
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.