Invitrogen
Manufacturer Code:PA5105311
Catalog # PIPA5105311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SKI around the phosphorylation site of Ser383. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310012I02Rik 2610001A11Rik AA062172 AA589460 BC004088 c-ski proto-oncogene c-Ski RGD1565591 SGS SKI ski oncogene ski oncoprotein SKI proto-oncogene ski sarcoma viral oncogene homolog ski sarcoma viral oncogene homolog (avian) SKV Sloan-Kettering viral oncogene homolog v-ski avian sarcoma viral oncogene homolog; C-ski; Proto-oncogene c-Ski; Ski oncogene; ski oncoprotein; ski sarcoma viral oncogene homolog; Sloan-Kettering viral oncogene homolog; v-ski avian sarcoma viral oncogene homolog
Gene Aliases: 2310012I02Rik; 2610001A11Rik; AA062172; AA589460; BC004088; c-ski; RGD1565591; SGS; SKI; SKV
UniProt ID: (Human) P12755
Entrez Gene ID: (Human) 6497, (Rat) 313757, (Mouse) 20481
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.