Invitrogen
Manufacturer Code:PA5105063
Catalog # PIPA5105063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SIN3A around the phosphorylation site of Ser832. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW553200 Histone deacetylase complex subunit Sin3a Kiaa4126 Mammalian Sin3A mKIAA4126 mSin3A paired amphipathic helix protein Sin3a SIN Sin3 SIN3 homolog A transcription regulator SIN3 transcription regulator family member A SIN3 transcription regulator homolog A SIN3A transcriptional corepressor Sin3a transcriptional co-repressor Sin3A transcriptional regulator SIN3 yeast homolog A transcriptional regulator SIN3A transcriptional regulator SIN3A (yeast); Histone deacetylase complex subunit Sin3a; Paired amphipathic helix protein Sin3a; SIN3 homolog A, transcription regulator; SIN3 transcription regulator homolog A; SIN3A; transcriptional co-repressor Sin3A; Transcriptional corepressor Sin3a; transcriptional regulator, SIN3 yeast homolog A; transcriptional regulator, SIN3A
Gene Aliases: AW553200; Kiaa4126; mKIAA4126; mSin3A; Sin3; SIN3A
UniProt ID: (Human) Q96ST3, (Mouse) Q60520
Entrez Gene ID: (Human) 25942, (Rat) 363067, (Mouse) 20466
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.