Invitrogen
Manufacturer Code:MA537377
Catalog # PIMA537377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Shp1Y536-2A7 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr536 of human phospho Shp1. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 2-8°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 70Z-SHP EC 3.1.3.48 hcp Hcph hematopoietic cell phosphatase Hematopoietic cell protein-tyrosine phosphatase HPTP1C me motheaten protein tyrosine phosphatase non-receptor type 6 protein-tyrosine phosphatase 1C Protein-tyrosine phosphatase SHP-1 PTN6 Ptp1C PTP-1C Ptph6 Ptpn6 PTPTY-42 SH2 phosphatase 1 SHP1 SHP-1 SHP-1L SH-PTP1 Tyrosine-protein phosphatase non-receptor type 6; hematopoietic cell phosphatase; Hematopoietic cell protein-tyrosine phosphatase; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTP-1C; SH-PTP1; Tyrosine-protein phosphatase non-receptor type 6
Gene Aliases: HCP; HCPH; HPTP1C; PTP-1C; PTP1C; PTPN6; SH-PTP1; SHP-1; SHP-1L; SHP1
UniProt ID: (Human) P29350
Entrez Gene ID: (Human) 5777
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.