Invitrogen
Manufacturer Code:PA5104855
Catalog # PIPA5104855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SHP-1 around the phosphorylation site of Tyr536. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 70Z-SHP; 70Z-SHP EC 3.1.3.48 hcp Hcph hematopoietic cell phosphatase Hematopoietic cell protein-tyrosine phosphatase HPTP1C me motheaten protein tyrosine phosphatase non-receptor type 6 protein-tyrosine phosphatase 1C Protein-tyrosine phosphatase SHP-1 PTN6 Ptp1C PTP-1C Ptph6 Ptpn6 PTPTY-42 SH2 phosphatase 1 SHP1 SHP-1 SHP-1L SH-PTP1 Tyrosine-protein phosphatase non-receptor type 6; hematopoietic cell phosphatase; Hematopoietic cell protein-tyrosine phosphatase; motheaten; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTP-1C; PTPTY-42; SH-PTP1; SH2 phosphatase 1; SHP-1; Tyrosine-protein phosphatase non-receptor type 6
Gene Aliases: 70Z-SHP; HCP; HCPH; HPTP1C; me; PTP-1C; PTP1C; Ptph6; PTPN6; PTPTY-42; SH-PTP1; SHP-1; SHP-1L; SHP1
UniProt ID: (Human) P29350, (Rat) P81718, (Mouse) P29351
Entrez Gene ID: (Human) 5777, (Rat) 116689, (Mouse) 15170
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.