Invitrogen
Manufacturer Code:PA517932
Catalog # PIPA517932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NSNSDpYVDVLP) corresponding to the internal amino acids of SH2D1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EAT-2; EWS/FLI1-activated transcript 2; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B; SH2D1B SH2 domain containing 1B EAT2 SH2 domain-containing molecule EAT2
Gene Aliases: EAT2; SH2D1B
UniProt ID: (Human) O14796
Entrez Gene ID: (Human) 117157
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.