Invitrogen
Manufacturer Code:MA515140
Catalog # PIMA515140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | G.22.6 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer235 and pSer236 of human ribosomal protein S6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S6; 40S ribosomal protein S6 OTTHUMP00000021121 OTTHUMP00000021122 phosphoprotein NP33; Phosphoprotein NP33; Small ribosomal subunit protein eS6
Gene Aliases: OK/SW-cl.2; RPS6; S6; S6R
UniProt ID: (Human) P62753, (Mouse) P62754, (Rat) P62755
Entrez Gene ID: (Human) 6194, (Mouse) 20104, (Rat) 29304
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.