Invitrogen
Manufacturer Code:PA538327
Catalog # PIPA538327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Rhodopsin around the phosphorylation site of Ser334 (D-A-SP-A-T) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L opsin; Long Wavelength Sensitive opsin; LWS opsin; opsin 2; opsin 2, rod pigment; Opsin-2; Red Opsin; RHO CSNBAD1 OPN2 Rhodopsin RP4 Opsin 2 rod pigment Opsin-2; Rhodopsin; Rhodopsin (retinitis pigmentosa 4, autosomal dominant); Rod Opsin
Gene Aliases: CSNBAD1; Noerg1; OPN2; Ops; RHO; RP4
UniProt ID: (Human) P08100, (Rat) P51489, (Mouse) P15409
Entrez Gene ID: (Human) 6010, (Rat) 24717, (Mouse) 212541
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.