Invitrogen
Manufacturer Code:PA5117194
Catalog # PIPA5117194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding S780 of human RB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cleaved Rb delta RB-p70 DRb-p70 exon 17 tumor GOS561 substitution mutation causes premature stop GOS563 exon 17 substitution mutation causes premature stop OSRC p104 P105RB p105-Rb Phospho-Retinoblastoma pp105 pp110 PPP1R130 pRb prepro-retinoblastoma-associated protein protein phosphatase 1 regulatory subunit 130 rb RB transcriptional corepressor 1 RB1 Rb-1 Rb-p70 Retin Retinoblastoma retinoblastoma 1 Retinoblastoma 1 (including osteosarcoma) retinoblastoma gene retinoblastoma protein retinoblastoma suspectibility protein retinoblastoma-associated protein; exon 17 tumor GOS561 substitution mutation causes premature stop; GOS563 exon 17 substitution mutation causes premature stop; p105-Rb; p110-RB1; pp105; pp110; pRb; prepro-retinoblastoma-associated protein; protein phosphatase 1, regulatory subunit 130; Rb; retinoblastoma 1; Retinoblastoma 1 (including osteosarcoma); retinoblastoma suspectibility protein; Retinoblastoma-associated protein
Gene Aliases: OSRC; p105-Rb; pp110; PPP1R130; pRb; RB; Rb-1; RB1
UniProt ID: (Human) P06400, (Rat) P33568, (Mouse) P13405
Entrez Gene ID: (Human) 5925, (Rat) 24708, (Mouse) 19645
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.