Invitrogen
Manufacturer Code:PA5105609
Catalog # PIPA5105609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AML1 around the phosphorylation site of Ser397. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acute myeloid leukemia 1 protein; acute myeloid leukemia 1 protein AML1 AML-1 aml1 oncogene AML1-EVI-1 AML1-EVI-1 fusion protein AMLCR1 CBF2alpha Cbfa2 CBF-alpha 2 CBF-alpha-2 core binding factor alpha 2 core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene) core-binding factor subunit alpha-2 core-binding factor runt domain alpha subunit 2 EVI-1 Oncogene AML-1 OTTHUMP00000108697 OTTHUMP00000108700 OTTHUMP00000108702 PEA2-alpha B Pebp2a2 PEBP2aB PEBP2alpha PEBP2-alpha B Pebpa2b Polyomavirus enhancer-binding protein 2 alpha B subunit RUN1 runt domain alpha subunit 2 runt related transcription factor 1 runt-related transcription factor 1 RUNX1 RUNX1/ZNF687 fusion SL3/AKV core-binding factor alpha B subunit SL3-3 enhancer factor 1 alpha B subunit transcription factor; AML1-EVI-1 fusion protein; CBF-alpha-2; core binding factor alpha 2; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); Core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; Oncogene AML-1; PEA2-alpha B; PEBP2-alpha B; Polyomavirus enhancer-binding protein 2 alpha B subunit; runt domain, alpha subunit 2; runt related transcription factor 1; Runt-related transcription factor 1; RUNX1/ZNF687 fusion; SL3-3 enhancer factor 1 alpha B subunit; SL3/AKV core-binding factor alpha B subunit; transcription factor
Gene Aliases: AML1; AML1-EVI-1; AMLCR1; CBF-alpha-2; CBF2alpha; CBFA2; EVI-1; Pebp2a2; PEBP2aB; PEBP2alpha; Pebpa2b; RUNX1
UniProt ID: (Human) Q01196, (Rat) Q63046
Entrez Gene ID: (Human) 861, (Rat) 50662, (Mouse) 12394
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.