Invitrogen
Manufacturer Code:PA538309
Catalog # PIPA538309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human P90RSK around the phosphorylation site of Thr359/Ser363 (TP-P-K-D-SP) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); HU-1 MAPKAPK1A RSK RSK1 90 kDa ribosomal protein S6 kinase 1 MAP kinase-activated protein kinase 1a MAPK-activated protein kinase 1a MAPKAP kinase 1a MAPKAPK-1a RSK-1 S6K-alpha 1 S6K-alpha-1 dJ590P13.1 (ribosomal protein S6 kinase 90kD polypeptide 1) p90-RSK 1 p90RSK1 p90S6K r HU-2 MAPKAPK1C RSK RSK3 S6K-alpha S6K-alpha2 p90-RSK3 pp90RSK3 90 kDa ribosomal protein S6 kinase 2 MAP kinase-activated protein kinase 1c MAPK-activated protein kinase 1c MAPKAP kinase 1c MAPKAPK-1c RSK-3 S6K-alpha-2 p90-RSK 2 p90RSK2 ribosomal S6 kinase 3 ribosomal protein S6 kinase alpha 2 ribosomal protein S6 kin RP11-393H10.3 CLS HU-3 ISPK-1 MAPKAPK1B MRX19 RSK RSK2 S6K-alpha3 p90-RSK2 pp90RSK2 90 kDa ribosomal protein S6 kinase 3 MAP kinase-activated protein kinase 1b; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1a; MAPKAP kinase 1a; MAPKAPK-1a; MAPKAPK1A; p90-RSK 1; p90RSK1; p90S6K; pp90RSK1; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; Ribosomal S6 kinase 1; RSK-1; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha-1
Gene Aliases: HU-1; Mapkapk-1a; MAPKAPK1A; p90-Rsk1; p90Rsk; p90Rsk1; p90S6K; RPS6KA1; RSK; Rsk-1; RSK1; S6K-alpha-1
UniProt ID: (Human) Q15418, (Rat) Q63531, (Mouse) P18653
Entrez Gene ID: (Human) 6195, (Rat) 81771, (Mouse) 20111
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.