Invitrogen
Manufacturer Code:PA538324
Catalog # PIPA538324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Rabphilin 3A around the phosphorylation site of Ser237 (R-A-SP-E-A) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exophilin-1; rabphilin 3A homolog; Rabphilin-3A; RPH3A rabphilin 3A homolog mouse
Gene Aliases: 2900002P20Rik; AU022689; AW108370; KIAA0985; RPH3A
UniProt ID: (Human) Q9Y2J0, (Rat) P47709, (Mouse) P47708
Entrez Gene ID: (Human) 22895, (Rat) 171039, (Mouse) 19894
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.