Invitrogen
Manufacturer Code:PA5105541
Catalog # PIPA5105541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Rpb1 CTD around the phosphorylation site of Ser2/5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 220kDa AW321876 DNA polymerase alpha 1 catalytic subunit DNA polymerase alpha 1 180 kDa DNA polymerase alpha 1 catalytic subunit DNA polymerase alpha catalytic subunit DNA polymerase alpha catalytic subunit p180 DNA polymerase alpha p180 subunit DNA polymerase alpha subunit I DNA polymerase epsilon catalytic subunit DNA polymerase epsilon catalytic subunit A DNA polymerase II subunit A DNA-directed RNA polymerase II largest subunit RNA polymerase II 220 kd subunit DNA-directed RNA polymerase II subunit A DNA-directed RNA polymerase II subunit RPB1 DNA-directed RNA polymerase III largest subunit DUN2 EC 2.7.7.6 G7a/Bat6Hom hRPB220 hsRPB1 I79_002751 I79_016081 MGC75453 N0825 NSX p180 Pol II S2p POL2 Pola Pola1 POLR2 Polr2a POLRA polymerase (DNA directed) alpha 1 polymerase (DNA directed) alpha 1 catalytic subunit polymerase (DNA) alpha 1 catalytic subunit polymerase (DNA-directed) alpha (70kD) Polymerase (RNA II (DNA directed) large polypeptide polymerase (RNA) II (DNA directed) polypeptide A polymerase (RNA) II (DNA directed) polypeptide A 220kDa polymerase (RNA) II subunit A POLYMERASE II polymerase alpha 1 RNA RNA POL2 RNA polymerase II 1 RNA polymerase II largest subunit RNA polymerase II subunit A RNA polymerase II subunit B1 RNA-directed RNA polymerase II subunit RPB1 RP02 RPB1 RPB220 RPBh1 Rpii215 RpIILS RPO2 RPO21 Rpo2-1 RPOL2 SUA8 YNL262W; DNA polymerase alpha 1 catalytic subunit; DNA polymerase alpha 1, 180 kDa; DNA polymerase alpha catalytic subunit; DNA polymerase alpha catalytic subunit p180; DNA polymerase alpha p180 subunit; DNA polymerase alpha subunit I; DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; G7a/Bat6Hom; polymerase (DNA directed), alpha 1, catalytic subunit; polymerase (DNA) alpha 1, catalytic subunit; polymerase (DNA-directed), alpha (70kD); Polymerase (RNA II (DNA directed), large polypeptide; polymerase (RNA) II (DNA directed) polypeptide A; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; polymerase, alpha 1; RNA polymerase II 1; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1
Gene Aliases: 220kDa; AW321876; hRPB220; hsRPB1; NSX; p180; POLA; POLA1; POLR2; POLR2A; POLRA; RPB1; RPBh1; Rpii215; RpIILS; RPO2; Rpo2-1; RPOL2
UniProt ID: (Human) P09884, (Human) P24928, (Mouse) P33609, (Rat) O89042, (Mouse) P08775
Entrez Gene ID: (Human) 5422, (Human) 5430, (Mouse) 18968, (Rat) 85241, (Mouse) 20020, (Rat) 363633
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.