Invitrogen
Manufacturer Code:PA538702
Catalog # PIPA538702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human RAD52 around the phosphorylation site of Tyr104 (K-F-YP-V-G) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD52 homolog; DNA repair protein RAD52 homolog recombination protein RAD52 rhabdomyosarcoma antigen MU-RMS-40.23; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23
Gene Aliases: RAD52; Rad52yh
UniProt ID: (Human) P43351, (Mouse) P43352
Entrez Gene ID: (Human) 5893, (Mouse) 19365
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.