Invitrogen
Manufacturer Code:PA537714
Catalog # PIPA537714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of serine 387 (R-I-S(p)-G-C) derived from Human GTPase Activating Protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYK4; GTPase activating protein GTPase activating protein ID-GAP RAC GTPase- activating protein 1 RACGAP1 RAC GTPase-activating protein GAP; Male germ cell RacGap; MgcRacGAP; Protein CYK4 homolog; Rac GTPase-activating protein 1; racgap1
Gene Aliases: AI227039; AI327394; Band25; CYK4; gtl11; GTPase; HsCYK-4; ID-GAP; KIAA1478; MGCRACGAP; mKIAA1478; RACGAP1
UniProt ID: (Human) Q9H0H5, (Mouse) Q9WVM1
Entrez Gene ID: (Human) 29127, (Mouse) 26934, (Rat) 315298
Molecular Function:
G-protein modulator
enzyme modulator
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.