Invitrogen
Manufacturer Code:MA537315
Catalog # PIMA537315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | PTENS380-NA9 |
Immunogen | A synthetic phospho peptide corresponding to residues surrounding Ser380 of human phospho PTEN. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 2-8°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 10q23del 2310035O07Rik A130070J02Rik AI463227 B430203M17Rik BZS CWS1 DEC GLM2 homolog of human mutated in multiple advanced cancers MHAM mitochondrial phosphatase and tensin protein alpha mitochondrial PTENalpha Mmac MMAC1 MMAC1 phosphatase and tensin homolog deleted on chromosome 10 Mutated in multiple advanced cancers 1 phosphatase and tensin homolog phosphatase and tensin homolog (mutated in multiple advanced cancers 1) phosphatase and tensin homolog deleted on chromosome ten phosphatase and tensin-like protein phosphatidylinositol 3 Phosphatidylinositol 345-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN phosphatidylinositol-3 phosphatidylinositol-345-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN protein tyrosine phosphatase and tensin-like protein Pten pten tumor suppressor PTEN1 TEP1; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; Mutated in multiple advanced cancers 1; Phosphatase and tensin homolog; phosphatase and tensin-like protein; Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN
Gene Aliases: 10q23del; 2310035O07Rik; A130070J02Rik; AI463227; B430203M17Rik; BZS; CWS1; DEC; GLM2; MHAM; MMAC1; PTEN; PTEN1; TEP1
UniProt ID: (Human) P60484, (Mouse) O08586
Entrez Gene ID: (Human) 5728, (Mouse) 19211
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.