Invitrogen
Manufacturer Code:PA5106230
Catalog # PIPA5106230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRPS1 around the phosphorylation site of S180. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310010D17Rik ARTS C76571 C76678 CMTX5 deafness 2 perceptive congenital deafness X-linked 2 perceptive congenital DFN2 DFNX1 dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1) phosphoribosyl pyrophosphate synthase I phosphoribosyl pyrophosphate synthetase 1 phosphoribosyl pyrophosphate synthetase I PPRibP PRPS1 Prps-1 PRSI PRS-I ribose-phosphate diphosphokinase 1 ribose-phosphate pyrophosphokinase 1; deafness 2, perceptive, congenital; deafness, X-linked 2, perceptive, congenital; dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1); Phosphoribosyl pyrophosphate synthase I; phosphoribosyl pyrophosphate synthetase I; PPRibP; PRS-I; ribose-phosphate diphosphokinase 1; Ribose-phosphate pyrophosphokinase 1
Gene Aliases: 2310010D17Rik; ARTS; C76571; C76678; CMTX5; DFN2; DFNX1; PPRibP; Prps-1; PRPS1; PRS-I; PRSI
UniProt ID: (Human) P60891, (Mouse) Q9D7G0, (Rat) P60892
Entrez Gene ID: (Human) 5631, (Mouse) 19139, (Rat) 29562
Molecular Function: carbohydrate kinase kinase ligase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.