Invitrogen
Manufacturer Code:PA537783
Catalog # PIPA537783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of serine 228(V-K-S(p)-N-N) derived from Human CARM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; Histone-arginine methyltransferase CARM1; PRMT4 histone-arginine methyltransferase CARM1 protein arginine N-methyltransferase 4; protein arginine methyltransferase; Protein arginine N-methyltransferase 4
Gene Aliases: CARM1; PRMT4
UniProt ID: (Human) Q86X55, (Mouse) Q9WVG6, (Rat) Q4AE70
Entrez Gene ID: (Human) 10498, (Mouse) 59035, (Rat) 363026
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.