Invitrogen
Manufacturer Code:PA5117190
Catalog # PIPA5117190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding S228 of human CARM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carm1 coactivator associated arginine methyltransferase 1 coactivator-associated arginine methyltransferase 1 coactivator-associated arginine methyltransferase 1 variant 5 histone-arginine methyltransferase CARM1 PRMT4 protein arginine methyltransferase protein arginine N-methyltransferase 4; Coactivator-associated arginine methyltransferase 1; Histone-arginine methyltransferase CARM1; Protein arginine N-methyltransferase 4
Gene Aliases: CARM1; PRMT4
UniProt ID: (Human) Q86X55
Entrez Gene ID: (Human) 10498
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.