Invitrogen
Manufacturer Code:PA5104884
Catalog # PIPA5104884
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PPAR- gamma around the phosphorylation site of Ser112. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CIMT1 GLM1 HUMPPARG LOC100008892 NR1C3 nuclear hormone receptor nuclear hormone receptor PPARG Nuclear receptor subfamily 1 group C member 3 OTTHUMP00000185032 OTTHUMP00000185036 peroxisome proliferative activated receptor gamma peroxisome proliferator peroxisome proliferator activated receptor peroxisome proliferator activated receptor gamma peroxisome proliferator activated receptor gamma 1 peroxisome proliferator activated receptor gamma 2 peroxisome proliferator activated receptor gamma 4 peroxisome proliferator activated receptor gamma-1 peroxisome proliferator activated receptor gamma-2 peroxisome proliferator activatived receptor gamma peroxisome proliferator activator receptor gamma peroxisome proliferator-activated nuclear receptor gamma variant 1 peroxisome proliferator-activated receptor gamma peroxisome proliferator-activated receptor gamma 1 peroxisome proliferator-activated receptor gamma 1a peroxisome proliferator-activated receptor gamma 1-a peroxisome proliferator-activated receptor gamma 1b peroxisome proliferator-activated receptor gamma 1-b peroxisome proliferator-activated receptor gamma 1c peroxisome proliferator-activated receptor gamma 1d peroxisome proliferator-activated receptor gamma 2 peroxisome proliferator-activated receptor-gamma peroxisome proliferator-activated receptor-gamma 2 peroxisome proliferator-cctivated receptor gamma 1 PPAR gamma PPAR gamma 1 PPAR gamma 3 PPARG PPARG1 PPARG2 PPARGAMMA PPAR-gamma PPARgamma2 PPAR-gamma2 PPARy proliferator activated gamma transcription factor; Nuclear receptor subfamily 1 group C member 3; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR-gamma
Gene Aliases: CIMT1; GLM1; NR1C3; PPAR-gamma; PPAR-gamma2; PPARG; PPARG1; PPARG2; PPARgamma; PPARgamma2
UniProt ID: (Human) P37231, (Mouse) P37238, (Rat) O88275
Entrez Gene ID: (Human) 5468, (Mouse) 19016, (Rat) 25664
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.