Invitrogen
Manufacturer Code:PA5110150
Catalog # PIPA5110150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding Y307 of human PPP2CA/PPP2CB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI115466 D8Ertd766e fb17h07 fc05c02 fk24a01 PP2A Pp2a1 Pp2a2 PP2A-alpha PP2Abeta PP2A-beta PP2Ac PP2CA PP2Calpha PP2CB PPP2CA PPP2CB R75353 Replication protein C RP C RP-C Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform wu:fb17h07 wu:fc05c02 wu:fk24a01 zgc:110641 zgc:55808 zgc:77172; PP2A-alpha; PP2A-beta; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase 2a, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; Replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146
Gene Aliases: AI115466; D8Ertd766e; PP2A; PP2Abeta; PP2Ac; PP2CA; PP2Calpha; PP2CB; PPP2CA; PPP2CB; R75353; RP-C
UniProt ID: (Human) P67775, (Human) P62714, (Mouse) P63330, (Mouse) P62715
Entrez Gene ID: (Human) 5515, (Human) 5516, (Mouse) 19052, (Mouse) 19053
Molecular Function:
calcium-binding protein
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.