Invitrogen
Manufacturer Code:PA5106201
Catalog # PIPA5106201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PNKP around the phosphorylation site of Ser114. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810009G08Rik 2'(3')-polynucleotidase AOA4 Bifunctional polynucleotide phosphatase/kinase DNA 5'-kinase/3'-phosphatase EIEE10 Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP) MCSZ phosphatase/kinase PKNP PNK PNKP Polynucleotide 3'-phosphatase Polynucleotide 5'-hydroxyl-kinase polynucleotide kinase 3'- phosphatase polynucleotide kinase 3'-phosphatase Polynucleotide kinase-3'-phosphatase; 2'(3')-polynucleotidase; Bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; Polynucleotide kinase-3'-phosphatase
Gene Aliases: AOA4; EIEE10; MCSZ; PNK; PNKP
UniProt ID: (Human) Q96T60
Entrez Gene ID: (Human) 11284
Molecular Function:
hydrolase
kinase
nucleotide kinase
nucleotide phosphatase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.