Invitrogen
Manufacturer Code:MA511124
Catalog # PIMA511124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 41AT858.235 |
Immunogen | KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S68 of human PLM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FXYD domain-containing ion transport regulator 1; Phospholemman; phospholemman FXYD1; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Aliases: 0610012C17Rik; 1110006M24Rik; FXYD1; PLM; Pml
UniProt ID: (Human) O00168, (Mouse) Q9Z239
Entrez Gene ID: (Human) 5348, (Mouse) 56188
Molecular Function: enzyme modulator ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.