Invitrogen
Manufacturer Code:PA538390
Catalog # PIPA538390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human PLD1 around the phosphorylation site of Ser561 (K-F-SP-L-Y) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline phosphatase 1; mPLD1; Phoshpolipase D gene 1 probably with two splicing variants A and B; Phosphatidylcholine-hydrolyzing phospholipase D1; phospholipase D gene 1, probably with two splicing variants A and B; Phospholipase D1; phospholipase D1, phosphatidylcholine-specific; PLD 1; PLD 1 choline phosphatase 1 hPLD1 phosphatidylcholine-hydrolyzing phospholipase D1 phospholipase D1 phospholipase D1 phophatidylcholine-specific; rPLD1
Gene Aliases: AA536939; C85393; PLD1; Pld1a; Pld1b; Plda; Pldb
UniProt ID: (Human) Q13393, (Rat) P70496
Entrez Gene ID: (Human) 5337, (Rat) 25096, (Mouse) 18805
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.