Invitrogen
Manufacturer Code:PA517790
Catalog # PIPA517790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer505 of human cPLA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium-dependent phospholipid-binding protein; cPLA2; cPLA2 calcium-dependent phospholipid-binding protein cytosolic phospholipase A2 lysophospholipase phosphatidylcholine 2-acylhydrolase phospholipase A2 group IVA; Cytosolic phospholipase A2; Lysophospholipase; Phosphatidylcholine 2-acylhydrolase; Phospholipase A2; phospholipase A2 cytosolic; Phospholipase A2 group IVA; phospholipase A2, group IVA (cytosolic, calcium-dependent)
Gene Aliases: CPLA2; cPLA2-alpha; cPLA2alpha; Pla2c; PLA2G4; PLA2G4A
UniProt ID: (Human) P47712, (Mouse) P47713
Entrez Gene ID: (Human) 5321, (Mouse) 18783, (Rat) 24653
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.