Invitrogen
Manufacturer Code:PA5104567
Catalog # PIPA5104567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PKD2 around the phosphorylation site of Ser812. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A MGC138466 APKD2 Autosomal dominant polycystic kidney disease type II protein C030034P18Rik MGC138468 PC2 Pc-2 PKD2 PKD4 polycystic kidney disease 2 polycystic kidney disease 2 (autosomal dominant) polycystic kidney disease 2 homolog Polycystic kidney disease 2 protein polycystic kidney disease 2 protein homolog polycystin 2 polycystin 2 transient receptor potential cation channel polycystin-2 Polycystwin R48321 RGD1559992 Transient receptor potential cation channel subfamily P member 2 transient receptor potential cation channel subfamily P member 2 TRPP1 TRPP2; Autosomal dominant polycystic kidney disease type II protein; PC2; polycystic kidney disease 2 (autosomal dominant); polycystic kidney disease 2 homolog; Polycystic kidney disease 2 protein; Polycystic kidney disease 2 protein homolog; polycystin 2; Polycystin-2; Polycystwin; R48321; Transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2
Gene Aliases: APKD2; C030034P18Rik; Pc-2; PC2; PKD2; PKD4; RGD1559992; TRPP2
UniProt ID: (Human) Q13563, (Mouse) O35245
Entrez Gene ID: (Human) 5311, (Rat) 498328, (Mouse) 18764
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.