Invitrogen
Manufacturer Code:PA5105195
Catalog # PIPA5105195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PKC delta around the phosphorylation site of Ser645. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-May AI385711 ALPS3 CVID9 D14Ertd420e kinase nPKC-delta KPCD MAY1 nPKC-delta PKC[d] PKCD PKCdelta Prkcd protein kinase C delta protein kinase C delta type Protein kinase C delta type catalytic subunit Protein kinase C delta type regulatory subunit protein kinase C delta variant IX protein kinase C delta VIII protein kinase C delta protein kinase C delta IV protein kinase C delta V protein kinase C[d] SDK1 Sphingosine-dependent protein kinase-1 Tyrosine-protein kinase PRKCD; nPKC-delta; Protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD
Gene Aliases: AI385711; ALPS3; CVID9; D14Ertd420e; MAY1; nPKC-delta; PKC[d]; PKCD; PKCdelta; PRKCD
UniProt ID: (Human) Q05655, (Mouse) P28867, (Rat) P09215
Entrez Gene ID: (Human) 5580, (Mouse) 18753, (Rat) 170538
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.