Invitrogen
Manufacturer Code:PA5105337
Catalog # PIPA5105337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PFKM around the phosphorylation site of Ser74. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-1-kinase; 6-phosphofructo-1-kinase 6-phosphofructokinase type A 6-phosphofructokinase muscle type AI131669 ATP-dependent 6-phosphofructokinase muscle type ATP-dependent 6-phosphofructokinase muscle type {ECO:0000255 ATP-PFK ATP-PFK {ECO:0000255 GSD7 HAMAP-Rule:MF_03184} PFK1 PFK-1 Pfk4 Pfk-4 PFKA PFK-A PFKM Pfk-m PFKX Phosphofructo-1-kinase isozyme A phosphofructokinase 1 phosphofructokinase 4 phosphofructokinase M variant 3/4 phosphofructokinase muscle phosphofructokinase muscle phosphofructokinase polypeptide X phosphofructokinase-1 A isozyme phosphofructokinase-A phosphofructokinase-M phosphofructokinase-M (connecting peptide) phosphohexokinase phosphohexokinase {ECO:0000255 PPP1R122 protein phosphatase 1 regulatory subunit 122; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type {ECO:0000255|HAMAP-Rule:MF_03184}; ATP-PFK; ATP-PFK {ECO:0000255|HAMAP-Rule:MF_03184}; PFK-A; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase 4; phosphofructokinase M variant 3/4; phosphofructokinase muscle; phosphofructokinase, polypeptide X; phosphofructokinase-1 A isozyme; phosphofructokinase-A; phosphofructokinase-M; phosphofructokinase-M (connecting peptide); Phosphohexokinase; phosphohexokinase {ECO:0000255|HAMAP-Rule:MF_03184}; protein phosphatase 1, regulatory subunit 122
Gene Aliases: AI131669; ATP-PFK; GSD7; PFK-1; Pfk-4; PFK-A; PFK-M; PFK1; Pfk4; PFKA; PFKM; PFKX; PPP1R122
UniProt ID: (Human) P08237, (Mouse) P47857, (Rat) P47858
Entrez Gene ID: (Human) 5213, (Mouse) 18642, (Rat) 65152
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.