Invitrogen
Manufacturer Code:PA5117211
Catalog # PIPA5117211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphorylated peptide around S483 of human PFKFB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4930568D07Rik 6PF-2-K/Fru-26-P2ase 2 6PF-2-K/Fru-26-P2ase heart-type isozyme 6-phosphofructo-2-kinase 6-phosphofructo-2-kinase/fructose-26-biphosphatase 2 6-phosphofructo-2-kinase/fructose-26-bisphosphatase 2 6-Phosphofructo-2-kinase/fructose-26-bisphosphatase 2 (heart) fructose 6-phosphate 2-kinase/fructose 26-bisphosphatase Fructose-26-bisphosphatase fructose-26-bisphosphatase cardiac isozyme PFK/FBPase 2 PFK-2 car PFK-2 cardiac PFK-2/FBPase-2 PFK-2/FBPase-2 gene B PFKFB cardiac Pfkfb2 RH2K; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2; 6PF-2-K/Fru-2,6-P2ase 2; 6PF-2-K/Fru-2,6-P2ASE heart-type isozyme; Fructose-2,6-bisphosphatase; fructose-2,6-bisphosphatase, cardiac isozyme; PFK/FBPase 2; PFKFB, cardiac
Gene Aliases: PFK-2/FBPase-2; PFKFB2
UniProt ID: (Human) O60825
Entrez Gene ID: (Human) 5208
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.