Invitrogen
Manufacturer Code:PA5114618
Catalog # PIPA5114618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDGF Receptor beta around the phosphorylation site of Tyr1021. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI528809 beta platelet-derived growth factor receptor Beta-type platelet-derived growth factor receptor CD140 antigen-like family member B CD140B CD140b antigen IBGC4 IMF1 JTK12 KOGS PDGF beta chain PDGF Receptor beta PDGFR Pdgfr1 PDGFR-1 PDGFRB PDGF-R-beta PDGFR-beta PENTT platelet derived growth factor receptor beta platelet derived growth factor receptor beta polypeptide platelet-derived growth factor receptor 1 Platelet-derived growth factor receptor beta platelet-derived growth factor receptor beta variant 1 Platelet-derived growth factor receptor beta platelet-derived growth factor receptor beta polypeptide; Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140b; PDGF beta chain; PDGF-R-beta; PDGFR-1; PDGFR-beta; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide
Gene Aliases: AI528809; CD140B; IBGC4; IMF1; JTK12; KOGS; PDGFR; PDGFR-1; PDGFR1; PDGFRB; PENTT
UniProt ID: (Human) P09619, (Rat) Q05030, (Mouse) P05622
Entrez Gene ID: (Human) 5159, (Rat) 24629, (Mouse) 18596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.