Invitrogen
Manufacturer Code:MA514905
Catalog # PIMA514905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | T.564.8 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr857 of human PDGF receptor ß |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); Alpha-type platelet-derived growth factor receptor; Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member A; CD140 antigen-like family member B; CD140a; CD140a antigen; CD140b; PDGF alpha chain; PDGF beta chain; PDGF-R-alpha; PDGF-R-beta; PDGFR-1; PDGFR-2; pdgfr-a; PDGFR-alpha; PDGFR-beta; PDGFRA/BCR fusion; Platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; platelet-derived growth factor receptor, alpha polypeptide; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein
Gene Aliases: AI115593; AI528809; APDGFR; CD140A; CD140B; GAS9; IBGC4; IMF1; JTK12; KOGS; PDGFACE; PDGFR; PDGFR-1; PDGFR-2; PDGFR1; PDGFR2; PDGFRA; PDGFRB; PENTT; RHEPDGFRA
UniProt ID: (Human) Q9UD28, (Human) P09619, (Rat) P20786, (Mouse) P26618, (Rat) Q05030, (Mouse) P05622
Entrez Gene ID: (Human) 5156, (Human) 5159, (Rat) 25267, (Mouse) 18595, (Rat) 24629, (Mouse) 18596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.