Invitrogen
Manufacturer Code:PA5105091
Catalog # PIPA5105091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PABP2 around the phosphorylation site of Ser150. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mPABII Nuclear poly(A)-binding protein 1 OPMD PAB2 PABII Pabp2 PABP-2 Pabp3 PABPN1 poly(A) binding protein 2 poly(A) binding protein II poly(A) binding protein nuclear 1 poly(A) binding protein nuclear 1 poly(A)-binding protein 2 poly(A)-binding protein II polyA binding protein II polyadenylate-binding nuclear protein 1 Polyadenylate-binding protein 2; Nuclear poly(A)-binding protein 1; PABII; PABP-2; poly(A) binding protein 2; poly(A) binding protein II; poly(A) binding protein, nuclear 1; poly(A)-binding protein 2; Poly(A)-binding protein II; Polyadenylate-binding nuclear protein 1; Polyadenylate-binding protein 2
Gene Aliases: mPABII; OPMD; PAB2; PABII; PABP-2; PABP2; Pabp3; PABPN1
UniProt ID: (Bovine) Q28165, (Human) Q86U42, (Mouse) Q8CCS6
Entrez Gene ID: (Bovine) 282298, (Human) 8106, (Rat) 116697, (Mouse) 54196
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.