Invitrogen
Manufacturer Code:PA513005
Catalog # PIPA513005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T310 of human OSR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: odd-skipped homolog; odd-skipped related 1; Protein odd-skipped-related 1; Protein odd-skipped-related 1 OSR1 ODD
Gene Aliases: ODD; OSR1
UniProt ID: (Human) Q8TAX0
Entrez Gene ID: (Human) 130497
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.