Invitrogen
Manufacturer Code:PA5105162
Catalog # PIPA5105162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KOR-1 around the phosphorylation site of Tyr369. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K-OR-1; kappa opioid receptor; kappa opioid receptor kappa-type opioid receptor KOPR1 KOR K-OR-1 KOR-1 MSL-1 Opiate receptor kappa-1 opioid receptor kappa 1 opioid receptor kappa 1 opioid receptor kappa 2 OPRK Oprk1 Oprk2 Ork1 R21 Ror-d; Kappa-type opioid receptor; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor, kappa 1; opioid receptor, kappa 2
Gene Aliases: K-OR-1; KOR; KOR-1; MSL-1; OPRK; OPRK1; Oprk2; R21; Ror-d
UniProt ID: (Human) P41145, (Mouse) P33534, (Rat) P34975
Entrez Gene ID: (Human) 4986, (Mouse) 18387, (Rat) 29335
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.