Invitrogen
Manufacturer Code:PA538252
Catalog # PIPA538252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Opioid Receptor sigma around the phosphorylation site of Ser363 (T-P-SP-D-G) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-OR-1; delta 1 opioid receptor; delta opioid receptor 1; Delta-type opioid receptor; DOR-1; K56; MSL-2; Opioid receptor A; opioid receptor, delta 1; OPRD1 D-OR-1 Delta-type opioid receptor Delta opioid receptor Delta opiate receptor Delta opioid receptor 1 Delta-1 opioid receptor Opioid delta receptor DOR-1 Opioid receptor delta 1 OPRD
Gene Aliases: DOR; DOR-1; mDOR; Nbor; OPRD; OPRD1; Ror-a
UniProt ID: (Human) P41143, (Mouse) P32300, (Rat) P33533
Entrez Gene ID: (Human) 4985, (Mouse) 18386, (Rat) 24613
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.