Invitrogen
Manufacturer Code:PA537657
Catalog # PIPA537657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of Serine 274(P-L-S(p)-P-P) derived from Human Neuro D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic helix-loop-helix factor 1; basic helix-loop-helix transcription factor; beta-cell E-box transactivator 2; Beta-cell E-box transcriptional activator 2; Beta2; BETA2 BHF-1 MODY6 NEUROD bHLHa3 basic helix-loop-helix transcription factor beta-cell E-box transactivator 2 class A basic helix-loop-helix protein 3 neurogenic differentiation factor 1 neurogenic helix-loop-helix protein NEUROD; BHF-1; bHLHa3; Class A basic helix-loop-helix protein 3; NeuroD; NeuroD1; neurogenic differentiation 1; Neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD
Gene Aliases: BETA2; BHF-1; BHLHA3; MODY6; NEUROD; NEUROD1
UniProt ID: (Human) Q13562, (Rat) Q64289, (Mouse) Q60867
Entrez Gene ID: (Human) 4760, (Rat) 29458, (Mouse) 18012
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.