Invitrogen
Manufacturer Code:PA5106018
Catalog # PIPA5106018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NFAT2 around the phosphorylation site of Ser172. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2210017P03Rik AI449492 AV076380 MGC138448 NFAT tra NFAT trans NFAT transcription complex cytosolic component NFAT2 NFATC NF-ATc NFATC1 NF-ATc1 NF-ATc1.2 Nfatcb nuclear factor of activated T cells cytoplasmic calcineurin dependent 1 nuclear factor of activated T-cells 1 nuclear factor of activated T-cells 'c' nuclear factor of activated T-cells cytoplasmic 1 nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 1 transcription factor NF-ATc; NF-ATc; NF-ATc1; NFAT transcription complex cytosolic component; nuclear factor of activated T-cells 'c'; Nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1; transcription factor NF-ATc
Gene Aliases: 2210017P03Rik; AI449492; AV076380; NF-ATC; NF-ATc1.2; NFAT2; NFATC; NFATC1; Nfatcb
UniProt ID: (Human) O95644
Entrez Gene ID: (Human) 4772, (Mouse) 18018
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.