Invitrogen
Manufacturer Code:PA5117228
Catalog # PIPA5117228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphorylated peptide around T853 of human PPP1R12A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200015F06Rik 130 kDa myosin-binding subunit of smooth muscle myosin phophatase 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase 130 kDa regulatory subunit of myosin phosphatase 133kDa myosin-binding subunit of smooth muscle myosin phosphatase (M133) 5730577I22Rik AA792106 AV099298 D10Ertd625e I79_016312 LOW QUALITY PROTEIN: protein phosphatase 1 regulatory subunit 12A M110 M130 M130 of smooth muscle myosin phosphatase MBS MBSP myosin binding subunit myosin phosphatase target subunit 1 myosin phosphatase target subunit 1 Myosin phosphatase-targeting subunit 1 myosin-binding subunit of myosin phosphatase mypt1 PP-1M PP1M subunit M110 PPP1R12A Protein phosphatase 1 regulatory subunit 12A protein phosphatase 1 regulatory (inhibitor) subunit 12A protein phosphatase 1 regulatory subunit 12A protein phosphatase myosin-binding subunit Protein phosphatase subunit 1M RCJMB04_22k13 Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110 si:dkey-28j4.1 smooth muscle myosin phosphatase myosin-binding subunit unm p82emcf unm_p82emcf zgc:110448 zgc:110448 protein; MBSP; myosin binding subunit; Myosin phosphatase target subunit 1; myosin phosphatase, target subunit 1; Myosin phosphatase-targeting subunit 1; PP-1M; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; Protein phosphatase myosin-binding subunit; Protein phosphatase subunit 1M; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; smooth muscle myosin phosphatase myosin-binding subunit
Gene Aliases: 1200015F06Rik; 5730577I22Rik; AA792106; AV099298; D10Ertd625e; M110; M130; MBS; MBSP; MYPT1; PPP1R12A
UniProt ID: (Human) O14974, (Rat) Q10728, (Mouse) Q9DBR7
Entrez Gene ID: (Human) 4659, (Rat) 116670, (Mouse) 17931
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.