Invitrogen
Manufacturer Code:PA5106099
Catalog # PIPA5106099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MLCK around the phosphorylation site of Tyr471. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 155 kda myosin light chain kinase homolog 9530072E15Rik A930019C19Rik AAT7 AW489456 Kinase-related protein KRP MLCK MLCK1 MLCK108 MLCK210 MSTP083 MYLK MYLK1 myosin light chain kinase myosin light chain kinase smooth muscle Myosin light chain kinase smooth muscle deglutamylated form myosin light chain kinase myosin light polypeptide kinase nmMlck smMLCK smooth muscle myosin light chain kinase smooth muscle myosin light chain kinase (61-kDa active fragment) telokin; Kinase-related protein; KRP; MLCK; Myosin light chain kinase, smooth muscle; Myosin light chain kinase, smooth muscle, deglutamylated form; myosin, light polypeptide kinase; smooth muscle myosin light chain kinase; Telokin
Gene Aliases: AAT7; KRP; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; MYLK; MYLK1; smMLCK
UniProt ID: (Human) Q15746
Entrez Gene ID: (Human) 4638
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.