Invitrogen
Manufacturer Code:MA536890
Catalog # PIMA536890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | MSK1T581-A5 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Thr581 of human phospho MSK1. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 3110005L17Rik 6330404E13Rik 90 kDa ribosomal protein S6 kinase 5 AI854034 kinase MSK1 KS6A5 mitogen and stress-activated protein kinase 1 MSK1 MSPK1 Nuclear mitogen- and stress-activated protein kinase 1 ribosomal protein S6 kinase A5 ribosomal protein S6 kinase alpha-5 ribosomal protein S6 kinase 90kDa polypeptide 5 ribosomal protein S6 kinase polypeptide 5 RLPK RLSK RPS6KA5 RSKL RSK-like protein kinase S6K-alpha-5; 90 kDa ribosomal protein S6 kinase 5; Nuclear mitogen- and stress-activated protein kinase 1; Ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kDa, polypeptide 5; RLSK; RSK-like protein kinase; RSKL; S6K-alpha-5
Gene Aliases: 3110005L17Rik; 6330404E13Rik; AI854034; MSK1; MSPK1; RLPK; RPS6KA5
UniProt ID: (Human) O75582, (Mouse) Q8C050
Entrez Gene ID: (Human) 9252, (Mouse) 73086
Molecular Function:
annexin
calcium-binding protein
calmodulin
cytoskeletal protein
intracellular calcium-sensing protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.