Invitrogen
Manufacturer Code:PA539795
Catalog # PIPA539795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human MKP-1/2 around the phosphorylation site of Ser296/318 (I-I-SP-P-N) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3CH134/CL100 PTPase; CL 100; Dual specificity protein phosphatase 1; Dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH1; Dual specificity protein phosphatase hVH2; MAP kinase phosphatase 1; MAP kinase phosphatase 2; MAP kinase phosphatase-1; mitogen-activated protein (MAP) kinase phosphatase-1; Mitogen-activated protein kinase phosphatase 1; Mitogen-activated protein kinase phosphatase 2; Mitogen-activated protein kinase phosphatase 2 MAP kinase phosphatase 2 MKP-2 Dual specificity protein phosphatase hVH2 DUSP-4 DUSP 4 MKPII MKP II Dual specificity protein phosphatase 1 MAP kinase phosphatase-1 MKP-1 Protein-tyrosine phosphatase CL100 Dual specificity protein phosphatase hVH1 CL100 MKP1 PTPN10 VH1; mitogen-activated protein kinase phosphatase-1; oxidative stress-inducible protein tyrosine phosphatase; protein tyrosine phosphatase non-receptor type 16; protein tyrosine phosphatase, non-receptor type 16; Protein-tyrosine phosphatase 3CH134; Protein-tyrosine phosphatase CL100; Protein-tyrosine phosphatase ERP; Protein-tyrosine phosphatase non-receptor type 16; serine/threonine specific protein phosphatase; VH1 homologous phosphatase 2
Gene Aliases: 2700078F24Rik; 3CH134; AI844617; BB104621; CL100; DUSP1; DUSP4; E130306H24Rik; erp; HVH1; HVH2; MKP-1; MKP-2; MKP1; MKP2; PTPN10; Ptpn16; TYP; U19515; VH1; VH2
UniProt ID: (Human) P28562, (Human) Q13115, (Rat) Q64623, (Mouse) P28563, (Rat) Q62767, (Mouse) Q8BFV3
Entrez Gene ID: (Human) 1843, (Human) 1846, (Rat) 114856, (Mouse) 19252, (Rat) 60587, (Mouse) 319520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.