Invitrogen
Manufacturer Code:PA537702
Catalog # PIPA537702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of serine 207 (V-D-S(p)-V-A) derived from Human MKK6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 6; MAPK/ERK kinase 6; MAPKK 6; MAPKK6 MEK6 MKK6 PRKMK6 SAPKK-3 SAPKK3 MAPK/ERK kinase 6 MAPKK 6 MEK 6 SAPK kinase 3 dual specificity mitogen-activated protein kinase kinase 6 protein kinase mitogen-activated kinase 6 (MAP kinase kinase 6) stress-activated protein kinase kinase 3 MAPKK3 MEK3 MKK3 PRKMK3 SAPKK-2 SAPKK2 MAP kinase kinase 3 MAPK/ERK kinase 3 MAPKK 3 MEK 3 SAPK kinase 2 dual specificity mitogen-activated protein kinase kinase 3 stress-activated protein kinase kinase 2; MEK 6; mitogen activated protein kinase kinase 6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 3; SAPKK3; Stress-activated protein kinase kinase 3
Gene Aliases: MAP2K6; MAPKK6; MEK6; MKK6; PRKMK6; SAPKK-3; SAPKK3; SKK3
UniProt ID: (Human) P52564, (Mouse) P70236
Entrez Gene ID: (Human) 5608, (Mouse) 26399, (Rat) 114495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.