Invitrogen
Manufacturer Code:MA537345
Catalog # PIMA537345
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | MEK12S217S221-H2 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Ser217/221 of human phospho MEK1/2. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AA589381 CFC3 CFC4 dual specificity mitogen activated protein kinase kinase 1 dual specificity mitogen activated protein kinase kinase 2 dual specificity mitogen-activated protein kinase kinase 1 Dual specificity mitogen-activated protein kinase kinase 2 EC 2.7.12.2 ERK activator kinase 1 ERK activator kinase 2 hypothetical protein LOC406728 kinase MEK1 MAP kinase kinase 1 MAP kinase kinase 2 MAP kinase/Erk kinase MAP kinase/Erk kinase 1 MAP2K1 MAP2K2 MAPK/ERK kinase 1 MAPK/ERK kinase 2 MAPKK 1 MAPKK 2 MAPKK1 MAPKK2 MEK 1 MEK 2 MEK1 MEK-1 MEK2 MEK-2 MEKK1 mitogen activated protein kinase kinase 1 mitogen activated protein kinase kinase 2 mitogen-activated protein kinase kinase 1 mitogen-activated protein kinase kinase 2 mitogen-activated protein kinase kinase 2 p45 mitogen-activated protein kinase kinase type 2 MK2 MKK1 MKK2 MP2K1 Prkmk1 Prkmk2 protein kinase mitogen activated kinase 1 p45 protein kinase mitogen activated kinase 2 p45 protein kinase mitogen-activated kinase 1 (MAP kinase kinase 1) RCJMB04_27f19 si:ch211-242m18.2 wu:fj56a12 wu:fj61b01 zgc:56557 zgc:56557 protein; Dual specificity mitogen-activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 1; ERK activator kinase 2; MAP kinase kinase 1; MAP kinase kinase 2; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MEK 1; MEK 2; mitogen-activated protein kinase kinase 2, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Gene Aliases: CFC3; CFC4; MAP2K1; MAP2K2; MAPKK1; MAPKK2; MEK1; MEK2; MKK1; MKK2; PRKMK1; PRKMK2
UniProt ID: (Human) Q02750, (Human) P36507
Entrez Gene ID: (Human) 5604, (Human) 5605
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.