Invitrogen
Manufacturer Code:MA515016
Catalog # PIMA515016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | C.158.9 |
Immunogen | Synthetic phosphopeptide corresponding to residues around pSer217/221 of human MEK1/2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; Diphospho-ERK; dp-ERK; dpERK; drosophila ERK; Dual specificity mitogen-activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 2; enhancer of seven in absentia 7; ERK; ERK activator kinase 1; ERK activator kinase 2; Erk MAP kinase; ERK/rolled; extracellular signal-regulated kinase; extracellular signal-regulated kinase A; Extracellular-regulated kinase A; Extracellular-signal-related kinase A; MAP kinase; MAP kinase kinase 1; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP kinase/Erk kinase 1; MAP-kinase; MAP2K1 Map2k2 ERK activator kinase 1 MAP kinase kinase 1 MAPK/ERK kinase 1 MAPKK 1 MEK 1 dual specificity mitogen-activated protein kinase kinase 1 mitogen-activated kinase 1 Dual specificity mitogen-activated protein kinase kinase 2 MAP kinase kinase 2 MAPKK 2 ERK activator kinase 2 MAPK/ERK kinase 2 MEK2 MKK2 PRKMK2; MAPK; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MEK 1; MEK 2; MEK2; mitogen activated protein kinase; mitogen activated protein kinase kinase 1; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase; Mitogen-activated protein kinase ERK-A; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 2, p45; mitogen-activated protein kinase kinase type 2; phospho-ERK; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen activated, kinase 2, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); Protein rolled; RE08694p; rl-PA; rl-PC; rl-PD; rl-PE; rl-PG; rl-PH; rl-PI; roll; sevenmaker
Gene Aliases: 12559; AA589381; BcDNA:RE08694; CFC3; CFC4; CG12559; CG18732; CT34260; CT39192; D-ERK; DERK; DERK-A; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmERK; DmERK-A; DmERKA; DmMAPK; dp-ERK; DpERK; dpERK1; dpMAPK; Dsor2; E(sina)7; EC2-1; EK2-1; ERK; ERK-A; Erk/Map kinase; Erk1; ERK1/2; ERKA; EY2-2; GroupII; l(2)41Ac; l(2R)EMS45-39; MAP-k; MAP2K1; MAP2K2; MAPK; MAPKK1; MAPKK2; MEK1; MEK2; MEKK1; MK2; MKK1; MKK2; p-ERK; pERK; pMAPK; PRKMK1; PRKMK2; RL; rl/ERK; rl/MAPK; rll; SEM; SR2-1; Su(Raf)2B
UniProt ID: (Human) Q02750, (Chicken) Q90891, (Human) P36507, (Mouse) P31938, (Rat) Q01986, (Rat) P36506, (Mouse) Q63932, (Fruit fly) P40417
Entrez Gene ID: (Chicken) 415549, (Human) 5604, (Chicken) 396349, (Human) 5605, (Mouse) 26395, (Rat) 170851, (Rat) 58960, (Mouse) 26396, (Fruit fly) 3354888
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.