Invitrogen
Manufacturer Code:PA5117184
Catalog # PIPA5117184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding serine 217/221 of human MAP2K1/MAP2K2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA589381 CFC3 CFC4 dual specificity mitogen activated protein kinase kinase 1 dual specificity mitogen activated protein kinase kinase 2 dual specificity mitogen-activated protein kinase kinase 1 Dual specificity mitogen-activated protein kinase kinase 2 EC 2.7.12.2 ERK activator kinase 1 ERK activator kinase 2 hypothetical protein LOC406728 kinase MEK1 MAP kinase kinase 1 MAP kinase kinase 2 MAP kinase/Erk kinase MAP kinase/Erk kinase 1 MAP2K1 MAP2K2 MAPK/ERK kinase 1 MAPK/ERK kinase 2 MAPKK 1 MAPKK 2 MAPKK1 MAPKK2 MEK 1 MEK 2 MEK1 MEK-1 MEK2 MEK-2 MEKK1 mitogen activated protein kinase kinase 1 mitogen activated protein kinase kinase 2 mitogen-activated protein kinase kinase 1 mitogen-activated protein kinase kinase 2 mitogen-activated protein kinase kinase 2 p45 mitogen-activated protein kinase kinase type 2 MK2 MKK1 MKK2 MP2K1 Prkmk1 Prkmk2 protein kinase mitogen activated kinase 1 p45 protein kinase mitogen activated kinase 2 p45 protein kinase mitogen-activated kinase 1 (MAP kinase kinase 1) RCJMB04_27f19 si:ch211-242m18.2 wu:fj56a12 wu:fj61b01 zgc:56557 zgc:56557 protein; Dual specificity mitogen-activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 1; ERK activator kinase 2; MAP kinase kinase 1; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP kinase/Erk kinase 1; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MEK 1; MEK 2; MEK2; mitogen activated protein kinase kinase 1; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 2, p45; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen activated, kinase 2, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Gene Aliases: AA589381; CFC3; CFC4; MAP2K1; MAP2K2; MAPKK1; MAPKK2; MEK1; MEK2; MEKK1; MK2; MKK1; MKK2; PRKMK1; PRKMK2
UniProt ID: (Human) Q02750, (Human) P36507, (Mouse) P31938, (Rat) Q01986, (Rat) P36506, (Mouse) Q63932
Entrez Gene ID: (Human) 5604, (Human) 5605, (Mouse) 26395, (Rat) 170851, (Rat) 58960, (Mouse) 26396
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.