Invitrogen
Manufacturer Code:PA5104953
Catalog # PIPA5104953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MEF2A around the phosphorylation site of Thr312. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A430079H05Rik ADCAD1 MADS box transcription enhancer factor 2 polypeptide A (myocyte enhancer factor 2A) mef2 MEF2A myocyte enhancer factor 2a myocyte-specific enhancer factor 2A RSRFC4 RSRFC9 Serum response factor-like protein 1; MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); Myocyte-specific enhancer factor 2A; Serum response factor-like protein 1
Gene Aliases: A430079H05Rik; ADCAD1; MEF2; MEF2A; RSRFC4; RSRFC9
UniProt ID: (Human) Q96D14, (Mouse) Q60929, (Rat) Q2MJT0
Entrez Gene ID: (Human) 4205, (Mouse) 17258, (Rat) 309957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.