Invitrogen
Manufacturer Code:PA5105319
Catalog # PIPA5105319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC16A1 around the phosphorylation site of Ser213. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL022710 FLJ36745 HHF7 MCT MCT 1 Mct1 MCT1D MGC44475 monocarboxylate transporter Monocarboxylate transporter 1 RATMCT1 RNMCT1 SLC16A1 SLC16A1 protein solute carrier 16 (monocarboxylic acid transporter) member 1 solute carrier family 16 (monocarboxylate transporter) member 1 solute carrier family 16 (monocarboxylic acid transporters) member 1 solute carrier family 16 member 1 solute carrier family 16 member 1 (monocarboxylic acid transporter 1); MCT 1; Monocarboxylate transporter 1; solute carrier 16 (monocarboxylic acid transporter), member 1; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; Solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Aliases: AL022710; HHF7; MCT; MCT1; MCT1D; RATMCT1; RNMCT1; SLC16A1
UniProt ID: (Human) Q9NSJ9, (Rat) P53987, (Mouse) P53986
Entrez Gene ID: (Human) 6566, (Rat) 25027, (Mouse) 20501
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.