Invitrogen
Manufacturer Code:PA564526
Catalog # PIPA564526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAP4 (Phospho-Ser696) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP 4; MAP-4; MAP-4 Microtubule-associated protein 4; Microtubule-associated protein 4
Gene Aliases: AA407148; MAP-4; MAP4; Mtap-4; Mtap4
UniProt ID: (Human) P27816, (Mouse) Q7TPD4, (Rat) Q5M7W5
Entrez Gene ID: (Human) 4134, (Mouse) 17758, (Rat) 367171
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.